siRNA Details
| Gene Symbol: SPATA5L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323640.1 | NP_001310569.1 | 5 | 1945 | ||
| NR_027635.1 | 5 | 1945 | ||||
| NR_136646.1 | 6 | 2140 | ||||
| NR_136647.1 | 6 | 2107 | ||||
| GenBank mRNA | AK023232.1 | - | 5 | 1891 | ||
| AK291457.1 | - | 5 | 1898 | |||

SPATA5L1
Genomic Map