siRNA Details
| Gene Symbol: LOC440040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027044.1 | 4 | 1341 | |||
| GenBank mRNA | AK294678.1 | - | 4 | 1341 | ||
| BC141819.1 | - | 4 | 1249 | |||
| BC142657.1 | - | 4 | 1249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440040| Gene Symbol: LOC440040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027044.1 | 4 | 1341 | |||
| GenBank mRNA | AK294678.1 | - | 4 | 1341 | ||
| BC141819.1 | - | 4 | 1249 | |||
| BC142657.1 | - | 4 | 1249 | |||