siRNA Details
| Gene Symbol: LOC284412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029390.1 | 1 | 391 | |||
| GenBank mRNA | BC045185.1 | - | 1 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284412| Gene Symbol: LOC284412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029390.1 | 1 | 391 | |||
| GenBank mRNA | BC045185.1 | - | 1 | 397 | ||