siRNA Details
| Gene Symbol: ST13P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002183.1 | 1 | 1249 | |||
| GenBank mRNA | AF539468.1 | - | 1 | 1249 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF279660.3 | - | 6 | 176224 | Chr. 13 - 49996975 - 50344526 [+] on Build GRCh38 | |

ST13P4
Genomic Map