siRNA Details
| Gene Symbol: FAM66C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026788.1 | 8 | 2197 | |||
| GenBank mRNA | BC048994.1 | - | 6 | 1554 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX394452.2 | - | 2 | 294 | Chr. 12 - 8196449 - 8200546 [+] on Build GRCh38 | |

FAM66C
Genomic Map