siRNA Details
Gene Symbol: ASZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028695.1 | - | 11 | 1953 | ||
BC034963.1 | - | 11 | 1959 | 4824818 | ||
BC068488.1 | - | 10 | 1844 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ASZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028695.1 | - | 11 | 1953 | ||
BC034963.1 | - | 11 | 1959 | 4824818 | ||
BC068488.1 | - | 10 | 1844 |