siRNA Details
| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028695.1 | - | 11 | 1331 | ||
| BC034963.1 | - | 11 | 1336 | 4824818 | ||
| BC068488.1 | - | 10 | 1222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASZ1| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028695.1 | - | 11 | 1331 | ||
| BC034963.1 | - | 11 | 1336 | 4824818 | ||
| BC068488.1 | - | 10 | 1222 | |||