siRNA Details
| Gene Symbol: LOC100128554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130748.1 | 11 | 2220 | |||
| GenBank mRNA | BC035235.1 | - | 12 | 2396 | ||
| BC039370.1 | - | 11 | 2220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128554| Gene Symbol: LOC100128554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130748.1 | 11 | 2220 | |||
| GenBank mRNA | BC035235.1 | - | 12 | 2396 | ||
| BC039370.1 | - | 11 | 2220 | |||