siRNA Details
| Gene Symbol: FAM86FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024254.1 | 5 | 2062 | |||
| Gene Symbol: FAM86EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130740.1 | 4 | 2732 | |||
| NR_130741.1 | 4 | 1786 | ||||
| NR_130742.1 | 4 | 1736 | ||||

FAM86FP
Genomic Map