siRNA Details
| Gene Symbol: ANO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030691.1 | 1 | 146 | |||
| XM_006718604.2 | XP_006718667.1 | 1 | 129 | |||
| XM_006718605.2 | XP_006718668.1 | 1 | 141 | |||
| GenBank mRNA | BC033036.2 | - | 1 | 102 | 4823645 | |
| DB450993.1 | - | 1 | 134 | |||

ANO1
Genomic Map