siRNA Details
| Gene Symbol: LINC00341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026779.1 | 1 | 405 | |||
| GenBank mRNA | BC008299.1 | - | 1 | 405 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AJ227888.1 | - | 1 | 48 | Chr. 3 - 16460061 - 16460370 [-] on Build GRCh38 | |

LINC00341
Genomic Map