siRNA Details
| Gene Symbol: TSPY26P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002781.1 | 1 | 806 | |||
| GenBank mRNA | BC101556.1 | - | 1 | 806 | ||
| BC101558.1 | - | 1 | 806 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ432315.1 | - | 1 | 329 | Chr. 20 - 32189136 - 32189883 [-] on Build GRCh38 | |

TSPY26P
Genomic Map