siRNA Details
| Gene Symbol: CSNK1A1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027320.1 | 1 | 705 | |||
| GenBank mRNA | BC028192.1 | - | 1 | 658 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI825331.1 | - | 1 | 294 | Chr. 15 - 36799633 - 36818095 [-] on Build GRCh38 | |

CSNK1A1P1
Genomic Map