siRNA Details
| Gene Symbol: CSNK1A1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027320.1 | 4 | 1132 | |||
| GenBank mRNA | BC028192.1 | - | 4 | 1085 | ||
| BC066972.1 | - | 4 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK1A1P1| Gene Symbol: CSNK1A1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027320.1 | 4 | 1132 | |||
| GenBank mRNA | BC028192.1 | - | 4 | 1085 | ||
| BC066972.1 | - | 4 | 923 | |||