siRNA Details
| Gene Symbol: PFN1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003242.3 | 1 | 980 | |||
| GenBank mRNA | AF345651.1 | - | 1 | 580 | ||
| BC137262.1 | - | 1 | 908 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PFN1P2| Gene Symbol: PFN1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003242.3 | 1 | 980 | |||
| GenBank mRNA | AF345651.1 | - | 1 | 580 | ||
| BC137262.1 | - | 1 | 908 | |||