siRNA Details
| Gene Symbol: HMGB3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002165.1 | 1 | 151 | |||
| GenBank mRNA | BC103849.1 | - | 1 | 151 | ||
| BC103850.1 | - | 1 | 151 | |||
| BC103851.1 | - | 1 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMGB3P1| Gene Symbol: HMGB3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002165.1 | 1 | 151 | |||
| GenBank mRNA | BC103849.1 | - | 1 | 151 | ||
| BC103850.1 | - | 1 | 151 | |||
| BC103851.1 | - | 1 | 151 | |||