siRNA Details
Gene Symbol: HMGB3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002165.1 | 1 | 565 | |||
GenBank mRNA | BC103849.1 | - | 1 | 565 | ||
BC103850.1 | - | 1 | 564 | |||
BC103851.1 | - | 1 | 564 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HMGB3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002165.1 | 1 | 565 | |||
GenBank mRNA | BC103849.1 | - | 1 | 565 | ||
BC103850.1 | - | 1 | 564 | |||
BC103851.1 | - | 1 | 564 |