siRNA Details
| Gene Symbol: LOC440173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027471.1 | 2 | 1902 | |||
| GenBank mRNA | BC027471.1 | - | 2 | 1900 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE736107.1 | - | 1 | 277 | Chr. 9 - 87008453 - 87008841 [-] on Build GRCh38 | |

LOC440173
Genomic Map