siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 10 | 905 | |||
| NR_110307.1 | 8 | 663 | ||||
| NR_110308.1 | 9 | 835 | ||||
| GenBank mRNA | BC033546.1 | - | 9 | 806 | ||
| BC070365.1 | - | 10 | 885 | |||
| BI829888.1 | - | 8 | 627 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI828372.1 | - | 8 | 655 | Chr. 14 - 66486655 - 66498424 [+] on Build GRCh38 | |

LINC00238
Genomic Map