siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 1, 2 | 193 | |||
| NR_110308.1 | 1, 2 | 193 | ||||
| GenBank mRNA | BC033546.1 | - | 1, 2 | 164 | ||
| BC056671.1 | - | 1, 2 | 197 | |||
| BC070365.1 | - | 1, 2 | 173 | |||

LINC00238
Genomic Map