siRNA Details
| Gene Symbol: SKINT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026749.2 | 7 | 975 | |||
| GenBank mRNA | BX118767.1 | - | 2 | 109 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | W32394.1 | - | 69 | Chr. - - [-] on Build | ||

SKINT1L
Genomic Map