siRNA Details
| Gene Symbol: ACVR2B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028389.1 | 2 | 1511 | |||
| GenBank mRNA | AK025619.1 | - | 2 | 1511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACVR2B-AS1| Gene Symbol: ACVR2B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028389.1 | 2 | 1511 | |||
| GenBank mRNA | AK025619.1 | - | 2 | 1511 | ||