siRNA Details
| Gene Symbol: PIK3CD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027045.1 | 2 | 1057 | |||
| GenBank mRNA | AK097325.1 | - | 2 | 1066 | ||
| BC016063.1 | - | 2 | 1036 | |||
| BC144613.1 | - | 1 | 1185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIK3CD-AS1| Gene Symbol: PIK3CD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027045.1 | 2 | 1057 | |||
| GenBank mRNA | AK097325.1 | - | 2 | 1066 | ||
| BC016063.1 | - | 2 | 1036 | |||
| BC144613.1 | - | 1 | 1185 | |||