siRNA Details
| Gene Symbol: SCARNA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003011.1 | 1 | 30 | |||
| GenBank mRNA | AJ609485.1 | - | 1 | 30 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX538250.1 | - | 2 | 5142 | Chr. 15 - 82750572 - 82757085 [+] on Build GRCh38 | |

SCARNA15
Genomic Map