siRNA Details
Gene Symbol: LINC00488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026767.1 | 3 | 2438 | |||
GenBank mRNA | AK123226.1 | - | 3 | 2438 | ||
CX865329.1 | - | 1 | 532 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026767.1 | 3 | 2438 | |||
GenBank mRNA | AK123226.1 | - | 3 | 2438 | ||
CX865329.1 | - | 1 | 532 |