siRNA Details
| Gene Symbol: LINC00488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026767.1 | 3 | 2260 | |||
| GenBank mRNA | AK123226.1 | - | 3 | 2260 | ||
| CX865329.1 | - | 1 | 354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00488| Gene Symbol: LINC00488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026767.1 | 3 | 2260 | |||
| GenBank mRNA | AK123226.1 | - | 3 | 2260 | ||
| CX865329.1 | - | 1 | 354 | |||