siRNA Details
Gene Symbol: LINC00488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026767.1 | 3 | 1058 | |||
GenBank mRNA | AK123226.1 | - | 3 | 1058 | ||
BC022563.1 | - | 3 | 1047 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000494582.1 | - | 3 | 1058 | Chr. 3 - 109178165 - 109185257 [+] on Build GRCh38 |
