siRNA Details
| Gene Symbol: SCARNA16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003013.1 | 1 | 149 | |||
| GenBank mRNA | AJ609486.1 | - | 1 | 149 | ||
| Gene Symbol: MIR6516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_106997.1 | 1 | 39 | |||

SCARNA16
Genomic Map