siRNA Details
| Gene Symbol: LOC284578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027086.1 | 1 | 2082 | |||
| GenBank mRNA | AK090710.1 | - | 1 | 1683 | ||
| BC043647.2 | - | 1 | 1150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284578| Gene Symbol: LOC284578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027086.1 | 1 | 2082 | |||
| GenBank mRNA | AK090710.1 | - | 1 | 1683 | ||
| BC043647.2 | - | 1 | 1150 | |||