siRNA Details
| Gene Symbol: SNORA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002990.1 | 1 | 71 | |||
| Gene Symbol: TBRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092132.1 | - | 1 | 1436 | ||

SNORA5B
Genomic Map