siRNA Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 2 | 63 | ||
| GenBank mRNA | BG718342.1 | - | 2 | 61 | ||
| DB444853.1 | - | 2 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO31| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282683.1 | NP_001269612.1 | 2 | 63 | ||
| GenBank mRNA | BG718342.1 | - | 2 | 61 | ||
| DB444853.1 | - | 2 | 64 | |||