siRNA Details
| Gene Symbol: SNORA29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002965.1 | 1 | 113 | |||
| GenBank mRNA | AJ609472.1 | - | 1 | 113 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA429995.1 | - | 1 | 251 | Chr. 6 - 159784784 - 159785871 [-] on Build GRCh38 | |

SNORA29
Genomic Map