siRNA Details
| Gene Symbol: UGT2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024010.1 | 3 | 944 | |||
| GenBank mRNA | AK293381.1 | - | 3 | 515 | ||
| BP262099.1 | - | 3 | 242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT2A3| Gene Symbol: UGT2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024010.1 | 3 | 944 | |||
| GenBank mRNA | AK293381.1 | - | 3 | 515 | ||
| BP262099.1 | - | 3 | 242 | |||