siRNA Details
| Gene Symbol: SNORA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002993.1 | 11 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORA35| Gene Symbol: SNORA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002993.1 | 11 | ||||