siRNA Details
| Gene Symbol: HAND2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003679.2 | 4 | 585 | |||
| NR_136192.1 | 4 | 3991 | ||||
| NR_136193.1 | 4 | 4688 | ||||
| NR_136194.1 | 4 | 4158 | ||||
| NR_136197.1 | 4 | 4062 | ||||
| NR_136198.1 | 3 | 3879 | ||||
| NR_136199.1 | 5 | 4201 | ||||
| NR_136200.1 | 4 | 4691 | ||||
| NR_136201.1 | 4 | 4858 | ||||
| GenBank mRNA | DQ831955.1 | - | 1 | 2751 | ||
| DQ831956.1 | - | 1 | 2751 | |||
| EF529495.1 | - | 4 | 2992 | |||
| EF529496.1 | - | 4 | 2915 | |||
| EF529497.1 | - | 4 | 2862 | |||
| EF529498.1 | - | 4 | 3127 | |||
| EF529499.1 | - | 4 | 492 | |||
| EF529502.1 | - | 3 | 380 | |||
| EF529503.1 | - | 4 | 659 | |||
| EF529504.1 | - | 5 | 701 | |||
| EF529505.1 | - | 4 | 585 | |||
| EU000853.1 | - | 4 | 3904 | |||
| EU000854.1 | - | 4 | 3904 | |||
| EU000855.1 | - | 3 | 4996 | |||
| EU000856.1 | - | 3 | 4996 | |||

HAND2-AS1
Genomic Map