siRNA Details
| Gene Symbol: HAND2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003679.2 | 4 | 445 | |||
| NR_136192.1 | 4 | 3851 | ||||
| NR_136193.1 | 4 | 4548 | ||||
| NR_136194.1 | 4 | 4018 | ||||
| NR_136197.1 | 4 | 3922 | ||||
| NR_136198.1 | 3 | 3739 | ||||
| NR_136199.1 | 5 | 4061 | ||||
| NR_136200.1 | 4 | 4551 | ||||
| NR_136201.1 | 4 | 4718 | ||||
| GenBank mRNA | DQ831955.1 | - | 1 | 2611 | ||
| DQ831956.1 | - | 1 | 2611 | |||
| EF529495.1 | - | 4 | 2852 | |||
| EF529496.1 | - | 4 | 2775 | |||
| EF529497.1 | - | 4 | 2722 | |||
| EF529498.1 | - | 4 | 2987 | |||
| EF529499.1 | - | 4 | 352 | |||
| EF529502.1 | - | 3 | 240 | |||
| EF529503.1 | - | 4 | 519 | |||
| EF529504.1 | - | 5 | 561 | |||
| EF529505.1 | - | 4 | 445 | |||
| EU000853.1 | - | 4 | 3764 | |||
| EU000854.1 | - | 4 | 3764 | |||
| EU000855.1 | - | 3 | 4856 | |||
| EU000856.1 | - | 3 | 4856 | |||

HAND2-AS1
Genomic Map