siRNA Details
| Gene Symbol: HAND2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003679.2 | 4 | 489 | |||
| NR_136192.1 | 4 | 3895 | ||||
| NR_136193.1 | 4 | 4592 | ||||
| NR_136194.1 | 4 | 4062 | ||||
| NR_136197.1 | 4 | 3966 | ||||
| NR_136198.1 | 3 | 3783 | ||||
| NR_136199.1 | 5 | 4105 | ||||
| NR_136200.1 | 4 | 4595 | ||||
| NR_136201.1 | 4 | 4762 | ||||
| GenBank mRNA | DQ831955.1 | - | 1 | 2655 | ||
| DQ831956.1 | - | 1 | 2655 | |||
| EF529495.1 | - | 4 | 2896 | |||
| EF529496.1 | - | 4 | 2819 | |||
| EF529497.1 | - | 4 | 2766 | |||
| EF529498.1 | - | 4 | 3031 | |||
| EF529499.1 | - | 4 | 396 | |||
| EF529502.1 | - | 3 | 284 | |||
| EF529503.1 | - | 4 | 563 | |||
| EF529504.1 | - | 5 | 605 | |||
| EF529505.1 | - | 4 | 489 | |||
| EU000853.1 | - | 4 | 3808 | |||
| EU000854.1 | - | 4 | 3808 | |||
| EU000855.1 | - | 3 | 4900 | |||
| EU000856.1 | - | 3 | 4900 | |||

HAND2-AS1
Genomic Map