siRNA Details
| Gene Symbol: SNORA73A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002907.2 | 1 | 167 | |||
| GenBank mRNA | CX783521.1 | - | 1 | 664 | ||
| L07382.1 | - | 1 | 168 | |||
| S63405.1 | - | 1 | 168 | |||
| Gene Symbol: SNHG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092726.1 | - | 1 | 1181 | ||

SNORA73A
Genomic Map