siRNA Details
| Gene Symbol: SNORA53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003015.1 | 1 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORA53| Gene Symbol: SNORA53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003015.1 | 1 | 166 | |||