siRNA Details
| Gene Symbol: SNORA54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002982.1 | 1 | 95 | |||
| GenBank mRNA | AJ609479.1 | - | 1 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORA54| Gene Symbol: SNORA54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002982.1 | 1 | 95 | |||
| GenBank mRNA | AJ609479.1 | - | 1 | 95 | ||