siRNA Details
| Gene Symbol: SNORA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002986.1 | 1 | 83 | |||
| GenBank mRNA | AJ609469.1 | - | 1 | 83 | ||
| AK091209.1 | - | 1 | 1616 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORA60| Gene Symbol: SNORA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002986.1 | 1 | 83 | |||
| GenBank mRNA | AJ609469.1 | - | 1 | 83 | ||
| AK091209.1 | - | 1 | 1616 | |||