siRNA Details
Gene Symbol: SNORA60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002986.1 | 11 | ||||
GenBank mRNA | AJ609469.1 | - | 11 | |||
AK091209.1 | - | 1 | 1544 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNORA60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002986.1 | 11 | ||||
GenBank mRNA | AJ609469.1 | - | 11 | |||
AK091209.1 | - | 1 | 1544 |