siRNA Details
| Gene Symbol: SNORA50C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002995.1 | 1 | 71 | |||
| Gene Symbol: SNHG25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CD050382.1 | - | 228 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD174478.1 | - | 1 | 206 | Chr. 17 - 64146223 - 64146476 [+] on Build GRCh38 | |

SNORA50C
Genomic Map