siRNA Details
| Gene Symbol: SNORA77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003019.1 | 1 | 22 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DW453250.1 | - | 1 | 41 | Chr. 1 - 203729576 - 203729749 [+] on Build GRCh38 | |

SNORA77
Genomic Map