siRNA Details
| Gene Symbol: LOC284632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027087.2 | 3 | 695 | |||
| GenBank mRNA | BC033556.1 | - | 3 | 695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284632| Gene Symbol: LOC284632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027087.2 | 3 | 695 | |||
| GenBank mRNA | BC033556.1 | - | 3 | 695 | ||