siRNA Details
| Gene Symbol: LINC00293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027012.1 | 8 | 626 | |||
| NR_027013.1 | 7 | 1173 | ||||
| GenBank mRNA | AK097475.1 | - | 8 | 626 | ||
| BC041354.1 | - | 7 | 1185 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR739932.1 | - | 6 | 487 | Chr. 8 - 46840895 - 46854211 [+] on Build GRCh38 | |

LINC00293
Genomic Map