siRNA Details
| Gene Symbol: LINC00293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027012.1 | 1 | 23 | |||
| NR_027013.1 | 1 | 23 | ||||
| GenBank mRNA | AK097475.1 | - | 1 | 23 | ||
| BC041354.1 | - | 1 | 35 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00293| Gene Symbol: LINC00293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027012.1 | 1 | 23 | |||
| NR_027013.1 | 1 | 23 | ||||
| GenBank mRNA | AK097475.1 | - | 1 | 23 | ||
| BC041354.1 | - | 1 | 35 | |||