siRNA Details
| Gene Symbol: LINC00115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024321.1 | 1 | 1140 | |||
| GenBank mRNA | AK026292.1 | - | 1 | 1125 | ||
| BC017762.1 | - | 1 | 1141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00115| Gene Symbol: LINC00115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024321.1 | 1 | 1140 | |||
| GenBank mRNA | AK026292.1 | - | 1 | 1125 | ||
| BC017762.1 | - | 1 | 1141 | |||