siRNA Details
| Gene Symbol: FLJ13224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026806.1 | 1 | 1180 | |||
| GenBank mRNA | AK023286.1 | - | 1 | 1180 | ||
| BC118557.1 | - | 1 | 1007 | |||
| BC118637.1 | - | 1 | 1007 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX107136.1 | - | 1 | 73 | Chr. 12 - 31325423 - 31325817 [+] on Build GRCh38 | |

FLJ13224
Genomic Map