siRNA Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027139.1 | 3 | 595 | |||
| GenBank mRNA | AK123122.1 | - | 3 | 659 | ||
| BC018073.1 | - | 3 | 539 | |||
| BC032490.1 | - | 3 | 549 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX375805.2 | - | 1 | 32 | Chr. X - 56976590 - 56984482 [-] on Build GRCh38 | |

SPIN3
Genomic Map